Learn More
Corning™ Costar™ Netwell™ Reagent Trays, Non-Sterile
GREENER_CHOICE
Specially designed Netwell™ carriers with handles allow simultaneous processing of up to 12 samples per carrier
Brand:  Corning™ 3517
 View more versions of this product
Product Code. 10621272
  • 261.90 € / Pack of 200
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Specially designed Netwell carriers and handles allow simultaneous processing of up to 12 samples per carrier
  • Polystyrene reagent trays are available in white for colorimetric reaction contrast, or black for better visibility of tissue sections
  • Each carrier kit contains eight carriers and eight handles
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tray
25/Pk.
200/Cs.
Black
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Costar™ Netwell™ Reagent Trays, Non-Sterile > Black