Learn More
Corning™ Polypropylene PCR Tubes with Attached Cap
GREENER_CHOICE
Manufactured from thin-walled polypropylene and designed for precise fit in heat blocks to optimize heat transfer. Corning™ PCR Tubes withstand forces up to 10,000 x g.
Brand:  Corning™ 6571
 View more versions of this product
Product Code. 10378611
  • 197.00 € / Pack of 960
Explore more special offers
Description
Description
Individual PCR tubes are made of thin wall polypropylene and designed for precise fit in heat blocks to optimize heat transfer. Tubes are RNase-/DNase-free, are autoclavable at 121°C, and withstand centrifugation to 10,000 x g.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNase, DNase-free
Clear
96/Pk.
Heat blocks for fast heat transfer
Non-sterile
10,000 x g
Autoclavable
Thin Wall PCR Tube
Clear
0.2 mL
Polypropylene
Conical bottom
Polypropylene
Flat Cap
Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Polypropylene PCR Tubes with Attached Cap > Clear; Thermowell; Flat; 0.2 mL