Learn More
Corning™ Snapwell™ Culture Inserts
Designed with a ring supported membrane for use with diffusion or Ussing chambers once cells are grown to confluence. Corning™ Snapwell™ Culture Inserts are primarily used for transport and electrophysiological studies.
Brand: Corning™ 3407
230.49 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
Descrizione
The Snapwell insert is a modified Transwell culture insert that contains a 12 mm diameter tissue culture-treated membrane supported by a detachable ring. The inserts are primarily used for transport and electrophysiological studies. Once cells are grown to confluence, this ring-supported membrane can be placed into either vertical or horizontal diffusion or Ussing chambers.
Order Info
Sterile culture inserts are packaged in 6-well cluster plates.
Specifica
6 inserts and 6-well plate | |
Polycarbonate | |
0.4 μm | |
12 mm | |
24/Cs. |
1.12 cm2 | |
1 x 108 pores/cm2 | |
Culture Insert | |
6/Pk. | |
Sterile |