missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Sterile Disposable Reagent Reservoirs
BIOPHARMAGREENER_CHOICE

Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.

Brand:  Corning™ 4872

302.77 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10712541

  • 340.83 € / Pack of 200
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Manufactured from modified polystyrene for strength. Corning™ Costar™ Sterile Disposable Reagent Reservoirs are ideal for repetitive filling of multichannel pipetters.

Costar™ reagent reservoirs are ideal for repetitively filling multichannel pipettors.

  • Manufactured from modified polystyrene (white)
  • Individually wrapped, sterile
  • Disposable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL
Polystyrene
35 mm
200/Cs.
Bulk
Single-use
145 x 85 x 35 mm
Sterile
145 mm
5/Pk.
85 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Costar™ Sterile Disposable Reagent Reservoirs > 100 mL; 5/Pk

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.