Learn More
Corning™ Costar™ Transtar™-96 Cartridge
Ideal for changing cell culture media and screening monoclonal antibodies
Brand:  Corning™ 7610
312.17 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
Product Code. 10127030
  • 328.60 € / Pack of 24
Estimated Shipment: 29-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
These are sterile replacement cartridges for the Transtar-96™ Well Liquid Transfer system.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pipetter Cartridge
Transtar-96 pipetter
1/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Costar™ Transtar™-96 Cartridge > Pipetter cartridge