missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miele Professional™ Mobile Injector, Module A 302
Module A 302 for the optimum loading of laboratory glassware.
Brand: Miele Professional™ 11116960
640.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Suitable for laboratory glassware such as Erlenmeyer flasks
- Equipped with 32 injector nozzles with plastic supports ID 110
- Nozzle spacing 57 mm across the width, 61 mm across the depth

Specifications
Injector Module | |
Laboratory Washer Baskets A 100 and A 150 | |
Four rows of eight injectors | |
132 x 215 x 479 mm |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Miele Professional™ Mobile Injector, Module A 302 > For Use With: Laboratory washer baskets A 100 and A 150
Spot an opportunity for improvement?Share a Content Correction