missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts
BIOPHARMAGREENER_CHOICE

Patented self-centering design provides independent access to both apical and basolateral membranes of the cell monolayer. Corning™ Transwell™Multiple Well Plate with Permeable Polycarbonate Membrane Inserts provide an ideal solution for studying anchorage-dependent and -independent cells.

Brand:  Corning™ 3401

414.51 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10567522

  • 436.33 € / Pack of 48
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

You can use cell culture inserts to:

  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings 10 μm thick translucent membrane
  • Pore sizes ranging from 0.4 μm to 8 μm diameters
  • Treated for optimal cell attachment
  • Supplied in multiple well plates
  • Membrane must be stained for cell visibility
  • Sterile
TRUSTED_SUSTAINABILITY
Specifications

Specifications

12 inserts/12 well plate
Polycarbonate
0.4 μm
Epithelial and endothelial cell culture; transport, migration/invasion, co-culture, and microbial pathogenesis studies
4 plates; 12 inserts/plate
Clear
Sterile
1.12 cm2
1.5 mL
Pre-loaded Microporous Insert in Tissue Culture Plate
12 mm
TC-Treated
TC-Treated
12
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Information

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number22224030 Certificate TypeCertificate of Quality Date01/04/2025 Catalog Number3401
Lot Number22124019 Certificate TypeCertificate of Quality Date22/03/2025 Catalog Number3401
Lot Number22024037 Certificate TypeCertificate of Quality Date05/03/2025 Catalog Number3401
Lot Number21824050 Certificate TypeCertificate of Quality Date22/02/2025 Catalog Number3401
Lot Number20924005 Certificate TypeCertificate of Quality Date08/02/2025 Catalog Number3401
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Corning™ Transwell™ Multiple Well Plate with Permeable Polycarbonate Membrane Inserts > 12 mm; 0.4 μm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.