Learn More
Corning™ Counting Chamber for Corning Cell Counter
Disposable counting chambers are composed of two 10 μL chambers. Designed to optimize cell counting throughput by enchaining speed and data accuracy in your day to day mammalian cell culture experiments.
Brand:  Corning™ 480202
205.82 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.
Product Code. 17740789
  • 222.00 € / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Disposable counting chambers are composed of two 10 μL chambers
  • Optimize cell counting throughput by enchaining speed and data accuracy in your day to day mammalian cell culture experiments
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Counting Chamber
Single-use
0.1 mm
2 Chambers
10 μL (approx.) per Chamber
1/Pk.
Corning Cell Counter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Corning™ Counting Chamber for Corning Cell Counter > Counting Chamber; Depth: 0.1 mm