Learn More
Tocris Bioscience™ Covidcil-19
Binds to SARS-CoV-2 frameshifting element and reduces frameshifting efficiency
Brand:  Tocris Bioscience™ 7485/50
565.86 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18746136
  • 638.00 € / 50mg
Explore more special offers
Description
Description
Covidcil-19 binds to the revised attenuator hairpin structure of the SARS-CoV-2 frameshifting element (FSE) with high affinity (Kd = 11 nM). It stabilizes the hairpin's folded state and reduces frameshifting efficiency in cells. Covidcil-19 inhibits viral propagation and reduces viral infectivity by > 3.5 orders of magnitude.
Specifications
Specifications
3-[[4-(Methylamino)-2-quinazolinyl]amino]benzoic acid
50 mg
C16H14N4O2
1225177-95-6
Antiviral Compounds
0.98