missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Covidcil-19

Binds to SARS-CoV-2 frameshifting element and reduces frameshifting efficiency

168.00 € - 638.00 €

Specifications

Chemical Name or Material 3-[[4-(Methylamino)-2-quinazolinyl]amino]benzoic acid
CAS 1225177-95-6
Target Antiviral Compounds
Molecular Formula C16H14N4O2
Purity 0.98
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18739256
View Documents
Tocris Bioscience™
7485/10
10 mg
168.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18746136
View Documents
Tocris Bioscience™
7485/50
50 mg
638.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Covidcil-19 binds to the revised attenuator hairpin structure of the SARS-CoV-2 frameshifting element (FSE) with high affinity (Kd = 11 nM). It stabilizes the hairpin's folded state and reduces frameshifting efficiency in cells. Covidcil-19 inhibits viral propagation and reduces viral infectivity by > 3.5 orders of magnitude.
Specifications

Specifications

3-[[4-(Methylamino)-2-quinazolinyl]amino]benzoic acid
Antiviral Compounds
0.98
1225177-95-6
C16H14N4O2
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications