missing translation for 'onlineSavingsMsg'
Learn More

CPC™ Plastic Quick-Disconnect Fittings

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables tests.

Brand:  CPC™ 31311-52

 View more versions of this product

Product Code. 17385589

  • 865.00 € / Pack of 250
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables

CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.

  • Material: Medical-grade ABS, polycarbonate and polysulfone
  • Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
  • Fow Rate: Cv∽5.5 (79.25) max.

  • Ergonomic thumb latch
  • Audible 'click' when connected
  • USP Class VI materials
  • Can be sterilized by autoclave, EtO, e-beam, or gamma
  • Coupling halves can be rotated when connected
  • Tapered barb
  • BSE/TSE free materials

  • Cell culture media packaging and transfer lines
  • Harvest and collection applications
  • Buffer solutions
  • Tube sets
  • Urology drainage/collection systems
  • Hypo/hyperthermia blankets

Specifications

Specifications

USP Class VI, ADCF
Coupling Insert
9.5 mm
1.3 in.
9.5mm
-40°F
15.2 mm
Silicone
60 psi
71°C
Hose Barb Non-Valved In-line Coupling Insert
Autoclavable
ABS
33 mm
Cell culture media packaging and transfer lines, harvest and collection applications
-40°C
0.6 in.
White
160°F
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers