Learn More
CPC™ Plastic Quick-Disconnect Fittings
Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables tests.
32.65 € - 865.00 €
Specifications
Certifications/Compliance | USP Class VI, ADCF |
---|---|
Autoclavable | Autoclavable |
For Use With (Application) | Cell culture media packaging and transfer lines, harvest and collection applications |
Min. Operating Temperature (English) | -40°F |
Min. Operating Temperature (Metric) | -40°C |
Product Code | Brand | Product Type | Material | Diameter (Metric) Inner | Color | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Material | Diameter (Metric) Inner | Color | Price | Quantity & Availability | ||
17385589
|
CPC™
31311-52 |
Hose Barb Non-Valved In-line Coupling Insert | ABS | 9.5 mm | White |
865.00 €
Pack of 250 |
In Stock
Log in to see stock. |
||
11780029
|
CPC™
31311-21 |
Hose Barb Non-Valved In-line Coupling Body | Polysulfone | 6.4 mm | Amber |
39.75 €
Pack of 5 |
Estimated Shipment: 17-12-2024
Log in to see stock. |
||
11739979
|
CPC™
31311-71 |
Hose Barb Non-valved Coupling Insert | Polysulfone | 6.4 mm | Amber |
32.95 €
Pack of 5 |
Estimated Shipment: 13-01-2025
Log in to see stock. |
||
11749979
|
CPC™
31311-72 |
Hose Barb Non-Valved In-line Coupling Insert | Polycarbonate | 9.5 mm | Amber |
32.65 €
Pack of 5 |
Estimated Shipment: 11-04-2025
Log in to see stock. |
||
11700629
|
CPC™
31311-22 |
Hose Barb Non-Valved In-line Coupling Body | Polysulfone | 9.5 mm | Amber |
38.55 €
Pack of 5 |
Estimated Shipment: 18-12-2024
Log in to see stock. |
||
Description
Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables
CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.
- Material: Medical-grade ABS, polycarbonate and polysulfone
- Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
- Fow Rate: Cv∽5.5 (79.25) max.
- Ergonomic thumb latch
- Audible 'click' when connected
- USP Class VI materials
- Can be sterilized by autoclave, EtO, e-beam, or gamma
- Coupling halves can be rotated when connected
- Tapered barb
- BSE/TSE free materials
- Cell culture media packaging and transfer lines
- Harvest and collection applications
- Buffer solutions
- Tube sets
- Urology drainage/collection systems
- Hypo/hyperthermia blankets
Specifications
USP Class VI, ADCF | |
Cell culture media packaging and transfer lines, harvest and collection applications | |
-40°C |
Autoclavable | |
-40°F |