missing translation for 'onlineSavingsMsg'
Learn More

CRISPR-Cas9 Antibody (6G12), Alexa Fluor™ 700, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Bio-Techne NBP2-52398AF700

Product Code. 18109539

  • 653.00 € / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CRISPR-Cas9
Monoclonal
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
Protein G purified
RUO
Primary
Bacteria
Purified
Western Blot, Immunocytochemistry, Immunofluorescence
6G12
Western Blot, Immunocytochemistry/Immunofluorescence
Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Recombinant C-terminal fragment of S.pyogenes CRISPR/Cas9. [UniProt# Q99ZW2]
0.1 mL
Gene Editing, Gene Knockouts, Gene Therapy
This CRISPR-Cas9 antibody (6G12) - C-terminus is specific to Cas9 from Streptococcus pyogene.
Store at 4C in the dark.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
CRISPR-Cas9 Antibody (6G12), Alexa Fluor™ 700, Novus Biologicals™ > 0.1mL; Alexa Fluor 700

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.