missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Cryo-Safe™ Cold Box
Made of molded polystyrene. Bel Art™ Cryo-Safe™ Cold Box. Has an internal 12-place support for 1.5 mL microcentrifuge tubes..
Brand: Bel-Art™ F18847-0002
This item is not returnable.
View return policy
Description
- When container is taken out of a -20°C (-4°F) freezer, it maintains a -15°C (-5°F) temperature for 1.5 hours, and will remain below 0°C (32°F) for four hours.
- Consists of a molded polystyrene container with cover
- Internal 12-place support for 0.05 oz. (1.5 mL) microcentrifuge tubes
- Space surrounding tubes is filled with a sealed-in material to maintain the low temperature required
Specifications
0.05 oz. | |
Safeguards enzymes and thermolabile substances from thermal degradation | |
Cryo-Safe Cold Box | |
1.5 mL | |
Not Autoclavable | |
-15°C |
Cover | |
Polystyrene | |
White | |
12 Microcentrifuge Tubes (15 mL) | |
No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bel-Art™ Cryo-Safe™ Cold Box > Internal 12 place support for 1.5mL microcentrifuge tubes
Spot an opportunity for improvement?Share a Content Correction