missing translation for 'onlineSavingsMsg'
Learn More

Taylor Wharton™ K Series Cryogenic Storage System 24K With CryoCon Automatic Level Controller

The high-capacity K Series systems accommodate various inventory control systems. They are connected to a liquid nitrogen tank by a hose and are equipped with a CryoCon automatic level controller which monitors and controls the level of liquid nitrogen.

Brand:  Taylor Wharton™ 24K+M505K

Product Code. 11763769

  • 25535.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Cryogenic System 24K

  • Vapour or liquid storage
  • Superior vacuum performance
  • Stainless steel vacuum vessel/durable powder coated cabinet
  • Castor mounted
  • High capacity - holds up to 38,350 x 2 mL vials, or up to 739,500 x 0.25 mL straws
  • Accommodates various inventory control systems
  • Hose connects the 24K to a liquid nitrogen tank
  • Suitable for storage of biological specimens or larger objects such as organs for transplantation

CryoCon Levelling System

  • Monitors and controls level of liquid nitrogen
  • Compliant to Medical Devices Directive MDD 93/42 EC
  • Level display and automatic refilling of nitrogen
  • Temperature display and control
  • Alarms for high temperature, level sensor fault, low nitrogen supply, open lid, and unauthorized access
  • Allows manual filling
  • Data logging with serial port for PC or printer
  • Simultaneous or sequential filling in the case of multiple installations
  • Connection for centralized gas bypass control
  • Automatic defogging when lid is open
  • Quick chill when lid is closed
  • Password controlled access and programming
  • Software updates can be forwarded by email
  • Available options include: RS 485 interface, Gas bypass (individual), 4-20mA, 0-2 and 0-10V interface for temperature recording, CryoData PC software, PC/Printer cable
Specifications

Specifications

Cryogenic storage system with CryoCon automatic level controller
24K
52 Days
Cryokonservator 24K
CryoCon controller
405.7 lb.
864 x 965 x 1118 mm
787 mm (neck)
7 L/Day
360 L
High capacity cryogenic storage
1 Ea.
184 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Taylor Wharton™ K Series Cryogenic Storage System 24K With CryoCon Automatic Level Controller > Capacity: 360 L

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.