Learn More
Thermo Scientific™ Cryopreservation Vials, Canes and Canisters
Maximize storage capacities in the laboratory by using these Cryopreservation Vials, Canes and Canisters.
Brand:  Thermo Scientific™ 4000217
 View more versions of this product
Product Code. 12750348
  • 5.23 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Maximize storage capacities using Thermo Scientific™ Cryopreservation Vials, Canes and Canisters.
Maximize your cryopreservation storage capacities with a range of vials, canes and canisters.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cryo cane, aluminum for 12.5mm diameter cryo tubes
10 mL
Cryotubes
Cryo Cane
Silver