missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cryopreservation Vials, Canes and Canisters

Maximize storage capacities in the laboratory by using these Cryopreservation Vials, Canes and Canisters.

5.23 € - 20.00 €


Products 4
Product Code Brand Type For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Type For Use With (Equipment) Price Quantity & Availability  
11874760
View Documents
Thermo Scientific™
4000211
Cryo Cane 6 Ampules
20.00 €
6 ampoules
In Stock
to see stock.
Add to basket
 
11884760
View Documents
Thermo Scientific™
4000214
Cryo Cane 2 Ampules
5.23 €
Each
Estimated Shipment: 18-08-2025
to see stock.
Add to basket
 
15422424
View Documents
Thermo Scientific™
4000216
Cryo Cane 9.2 and 10 mL Goblets
5.63 €
Each
Estimated Shipment: 02-09-2025
to see stock.
Add to basket
 
12750348
View Documents
Thermo Scientific™
4000217
Cryo Cane 12.5 mm dia. Cryo Tubes
5.23 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Maximize storage capacities using Thermo Scientific™ Cryopreservation Vials, Canes and Canisters.

  • Warranty varies by country, please contact us for details
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Cryopreservation Vials, Canes and Canisters

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.