missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Cryotome™ Cryostat Cryocassettes, 25mm dia. Crosshatched
Enhance operation of Cryotome™ FSE/FE/SME/E Cryostats with Epredia™ Cryotome™ Cryostat Accessories.
Brand: Epredia™ A78930166
This item is not returnable.
View return policy
Description
- Multipositional heat extractor is incorporated on a vertical shaft for even freezing and easy removal from specimen; can be operated from any position on the freeze bar
- Freezing time is shortened if freezing stage and tissue are placed on active Peltier
- Freezing disc is used in conjunction with heat extractor, assisting in the flattening and freezing of tissue samples
Specifications
Narrow shelf (56mm) | |
Narrow Shelf | |
FSE/FE/SME/E |