Learn More
CleanSpace™ Technology CST Half Mask with Head Harness
A CleanSpace CST Half Masks (including a CleanSpace CST Head Harness) is designed for use with the CleanSpace PRO Power Unit (CST1000) and CleanSpace ULTRA Power Unit (CST 1010).
Brand:  CleanSpace™ Technology CST1036
 View more versions of this product
Product Code. 17835085
  • 198.00 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed for use with the CleanSpace PRO Power Unit (CST1000) and CleanSpace ULTRA Power Unit (CST 1010).
  • Made of soft silicone that readily moulds to the face and does not cause allergic dermatitis.
  • The easy-to-locate clips secure the CleanSpace CST Half Mask to the Power Unit.
  • Attaches easily to connection points on the mask arms and can be removed for cleaning.
  • A CleanSpace CST Half Mask (including a CleanSpace CST Head Harness) is NOT certified intrinsically safe.
Specifications
Specifications
Black
Silicon
Large/X-Large
Reusable
1/2 Mask and Harness