Learn More
CleanSpace™ Technology CST Neck Support
CleanSpace CST Neck Supports are specifically designed to provide additional positioning options of the power unit for optimal wearer comfort.
Brand:  CleanSpace™ Technology CST1020
 View more versions of this product
Product Code. 17805095
  • 70.65 € / Each
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • CleanSpace CST Neck Supports are specifically designed to provide additional positioning options of the power unit for optimal wearer comfort.
  • Compatible with the CleanSpace PRO Power Unit (CST1002) and CleanSpace ULTRA Power Unit (CST1012).
  • Made of light weight polypropylene plastic.
  • Clips directly into the Power Unit and is easily changed.
  • A CleanSpace CST Neck Support is NOT certified intrinsically safe.
Specifications
Specifications
Black
Polypropylene
Medium
Reusable
Neck Support