Learn More
Gibco™ CTS™ RevitaCell™ Supplement (100X)
CTS RevitaCell Supplement (100X) is a cGMP-manufactured, animal origin-free, chemically defined supplement that is used with pluripotent stem cells (PSCs), either as a post-thaw recovery solution to improve cell viability or in combination with CTS Essential 8 Medium to enhance single cell passaging applications.
Brand: Gibco™ A4238401
Description
CTS RevitaCell Supplement (100X) is a cGMP-manufactured, animal origin-free, chemically defined supplement that is used with pluripotent stem cells (PSCs), either as a post-thaw recovery solution to improve cell viability or in combination with CTS Essential 8 Medium to enhance single cell passaging applications. CTS RevitaCell Supplement reduces the burden in qualifying reagents when transitioning from the research bench to the clinic.
CTS RevitaCell Supplement:
• Facilitates regulatory filings through cGMP manufacturing and the availability of documentation
• Simplifies the transition from bench to clinic through the direct replacement of RUO RevitaCell Supplement
Facilitates regulatory filings
CTS RevitaCell Supplement is manufactured at a site that uses methods and controls that conform with cGMP for medical devices, 21 CFR Part 820. Our FDA-registered manufacturing site is ISO 13485-certified. Full documentation traceability and convenient access to our Drug Master File (DMF) are available for CTS RevitaCell Supplement.
Simplifies transition from bench to clinic
Like the RUO RevitaCell Supplement, the CTS RevitaCell Supplement maximizes cell viability post-single cell passaging and yields high post-thaw cell viability and growth of pluripotent stem cells. Both the RUO and CTS RevitaCell supplements include a ROCK inhibitor that has higher specificity than the traditional inhibitors, coupled with molecules containing antioxidant and free radical scavenger properties that minimize the impact of stress on the cells. With its additional safety testing, including adventitious agents, CTS RevitaCell Supplement enables a smooth transition from RUO RevitaCell Supplement.

Specifications
Store at -5 to -20°C and protect from light. |
|
Animal Origin-free, Chemically-defined | |
Liquid | |
Sterile-filtered | |
ISO 13485, MDSAP, FDA-registered, 21 CFR 820 | |
Supplied at a concentration of 0.9 mg/mL |
Pluripotent Stem Cells (PSCs) | |
100X | |
Supplement | |
Cell and Gene Therapy Research, Development and Manufacturing | |
5 mL | |
Dry Ice |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2366881 | Certificate of Analysis | 29/01/2025 | A4238401 |
2449298 | Certificate of Analysis | 29/01/2025 | A4238401 |
2524627 | Certificate of Analysis | 29/01/2025 | A4238401 |
2753926 | Certificate of Analysis | 29/01/2025 | A4238401 |
2824820 | Certificate of Analysis | 28/01/2025 | A4238401 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.