Learn More
Thermo Scientific™ Culti-Loops™ Kocuria rhizophila ATCC® 533™
Culti-Loops are disposable bacteriological loops containing stabilised viable microorganisms.
Brand: Thermo Scientific™ R4604070
Description
Use Thermo Scientific™ Culti-Loops™ Kocuria rhizophila ATCC™ 533 in the performance testing of culture media, stains, diagnostic kits, and reagents, for the maintenance of stock cultures and in the evaluation of bacteriological procedures. Culti-Loops are ready to use, disposable bacteriological loops containing stabilised viable microorganisms.
The film in each loop is made from a gelatine formulation and then dried by special processing. To rehydrate the film, the loops must come into contact with both warmth and moisture. Most organisms grow in 24–48 hours under the proper conditions. However, some exhibit a considerable lag phase and should be incubated for an additional 24 hours.
- Ready-to-use – Prepared disposable bacteriological loops with viable microorganisms
- Easy-to-store – Store at 2-8°C
There are two methods such as direct streak method and indirect streak method which may be used for inoculation. The appropriate method is used for the selected microorganism.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
533TM* | |
Kocuria rhizophila | |
Single Organism | |
Qualitative QC | |
5 Loops/Pk. |
Culti-Loops are CE Marked. | |
Kocuria rhizophila | |
Cocci | |
5 Loops/Pack |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
693912 | Certificate of Analysis | 16/04/2024 | R4604070 |
680741 | Certificate of Analysis | 16/04/2024 | R4604070 |
263052 | Certificate of Analysis | 16/04/2024 | R4604070 |
094082 | Certificate of Analysis | 16/04/2024 | R4604070 |
216816 | Certificate of Analysis | 03/04/2024 | R4604070 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.