missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiCHO™ Expression Medium

A GMP-manufactured, chemically-defined, serum-free, protein-free, animal origin–free medium specifically designed for high-density growth and transfection of suspension-adapted Chinese hamster ovary (CHO) cells.

Brand:  Gibco™ A2910003

 View more versions of this product

Product Code. 15461044

  • 4895.00 € / 10 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

ExpiCHO Expression Medium is specifically designed for high-density growth and transfection of suspension-adapted Chinese hamster ovary (CHO) cells. This medium is GMP-manufactured, chemically-defined, serum-free, protein-free, and animal origin–free. It is a core component of the ExpiCHO Expression System Kit (Cat. No. A29133) for scalable transient protein production. To achieve the highest levels of transient protein expression, the medium is matched to a culture feed, ExpiCHO Feed, also included in the system kit and transfection kits.

With ExpiCHO Expression Medium you can:

  • Grow suspension CHO cultures to densities of over 2 x 107 cells per mL, with >95% cell viability
  • Achieve high transfection efficiencies of >90% when coupled with ExpiFectamine CHO transfection reagent
  • Transiently transfect suspension CHO cells at a density of 6 x 106 cells per mL
  • Achieve yields up to 3 g per L of recombinant protein of high-density transiently transfected cultures by sustained, high-level expression
  • Grow and transfect CHO cells at volumes of <1 mL up to 5 L in culture formats from multiwell plates up to 5 L shake flask

ExpiCHO Expression Medium is ready to use. It is formulated with GlutaMAX-I reagent and does not require supplementation. It does not contain phenol red. The chemically-defined, animal origin free formulation results in high reproducibility and lot-to-lot reliability. ExpiCHO Expression Medium is not recommended for adherent cell cultures.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: in the dark at 2-8°C
Shipping conditions: ambient
Shelf life: 12 months from date of manufacture
Animal Origin-free, Chemically-defined, Protein-free, Serum-free
Liquid
Serum-free
No Phenol Red
ExpiCHO™
Room Temperature
ExpiCHO-S™, CHO-S, SFM Adapted
Suspension Cell Culture
CHO Expression Medium
GlutaMAX
Universal Bag
10 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.