missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Curved Gamma Radiation Shields

15° angled hour-glass shape shield ideal for providing clear non-distorted vision and reducing light images and shadows.

Brand:  Fisherbrand™ CSR-S4TG

Product Code. 12317197

  • 2800.00 € / Each
Estimated Shipment: 21-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Gamma – Hourglass Shield – Curved Base 30 x 45cm
  • 15° angled hour-glass shape shield provides clear non-distorted vision, reducing light images and shadows
  • The curved sides provide the user with easier access to the central area behind the shield
  • Available with either a large flat base to provide bench protection, or a curved base which fits conveniently over safety trays
Specifications

Specifications

15° Angle
Gamma Radiation Shield
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Curved Gamma Radiation Shields > Angle: 15°

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.