missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Custom mirVana™ miRNA Inhibitor

miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically inhibit endogenous miRNAs for the purpose of functional analysis via down-regulation.

Brand:  Invitrogen™ 4464087

915.22 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 15591124

  • 1366.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Use our GeneAssist™ miRNA Workflow Builder to design mimics and inhibitors for any miRNA sequence.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5°C to -30°C).
miRNA
miRNA
20 nmol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.