missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Custom mirVana™ miRNA Inhibitor
miRNA inhibitors are small, chemically modified single-stranded RNA molecules designed to specifically inhibit endogenous miRNAs for the purpose of functional analysis via down-regulation.
1366.00 €
Specifications
Content And Storage | Store in freezer (-5°C to -30°C). |
---|---|
RNAi Type | miRNA |
Product Type | miRNA |
Description
Use our GeneAssist™ miRNA Workflow Builder to design mimics and inhibitors for any miRNA sequence.
Specifications
Store in freezer (-5°C to -30°C). | |
miRNA | |
miRNA |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title