Learn More
Ambion™ Custom RNA, Stealth
Custom RNA, Stealth
Brand: Ambion™ 100001866
158.79 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
Use this tool for fast and easy online ordering of bench-tested Validated Stealth RNAi™ siRNA, ready-to-order Stealth Select RNAi™ siRNA, bench-tested BLOCK-iT™ Pol II miR Validated miRNA Vector Duopaks, and ready-to-clone BLOCK-iT™ miR RNAi Select.
Specifications
Room temperature | |
Stealth RNAI In-vivo, 20nmol, Lowest Off-target Effects and Highest Potency, Small, Chemically Modified Single-Stranded RNA Molecules Designed to Specifically Inhibit Endogenous miRNAs for the Purpose of Functional Analysis Via Down-Regulation | |
Stealth RNAI In-vivo | |
20 nmol |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Product Title
Ambion™ Custom RNA, Stealth > 20 nmol
Spot an opportunity for improvement?Share a Content Correction