missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ambion™ Custom Select siRNA
Custom Select siRNA
Brand: Ambion™ 4390827
216.41 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.Specifications
Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °C | |
siRNA | |
RNAi, epigenetics and non-coding RNA research, RNAi, siRNA | |
Custom Select siRNA | |
5nmol |
Custom Select siRNA, 5nmol, For RNAi, Epigenetics and Non-Coding RNA research, RNAi, siRNA | |
Custom select siRNA | |
Silencer™, Ambion™ | |
5 nmol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction