Learn More
Ambion™ Custom Select siRNA
Custom Select siRNA
323.00 € - 16270.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15531044
|
Ambion™
4390827 |
5 nmol |
323.00 €
5nmol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15551044
|
Ambion™
4390829 |
40 nmol |
570.00 €
40nmol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15541044
|
Ambion™
4390828 |
20 nmol |
492.00 €
20nmol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15561044
|
Ambion™
4390830 |
20 nmol |
682.00 €
20nmol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15571044
|
Ambion™
4390831 |
40 nmol |
774.00 €
40nmol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15561064
|
Ambion™
4404013 |
250 nmol |
2405.00 €
250nmol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15571084
|
Ambion™
4444245 |
1 μmol |
4105.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15561084
|
Ambion™
4444207 |
10 μmol |
16270.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.For Research Use Only. Not for use in diagnostic procedures.