Learn More
Ambion™ Custom Select siRNA
Custom Select siRNA
Brand: Ambion™ 4444207
This item is not returnable.
View return policy
Description
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.Specifications
Contains dried siRNA and Nuclease-free Water (1.75 mL), shipped at ambient temperature. Upon receipt, store siRNAs in a non-frost-free freezer at or below –20 °C | |
Silencer™ Select, Ambion™ | |
HPLC |
siRNA | |
siRNA | |
10 μmol |