Learn More
Ambion™ Custom Select siRNA
Custom Select siRNA
Brand: Ambion™ 4390831
518.58 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Silencer™ Select custom siRNAs (LNA-modified 21-bp duplex with overhangs) are RNA interference (RNAi) tools ideal for knocking down gene expression in order to study protein function in a wide range of cell types. These siRNAs provide high knock-down efficiencies and specificities.Specifications
Room temperature | |
Knocking down gene expression in order to study protein function in a wide range of cell types | |
HPLC |
Custom Select siRNA, 40nmol, HPLC Purity, ≥95% Purity, Ideal for Knocking Down Gene Expression in Order to Study Protein Function in a Wide Range of Cell Types, These siRNAs Provide High Knock-down Efficiencies and Specificities | |
Custom Select siRNA | |
40 nmol |