Learn More
Ambion™ Custom Silencer™ siRNA
Custom Silencer™ siRNA
Brand: Ambion™ AM16211
391.28 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.Specifications
Room temperature | |
Custom siRNA | |
20 nmol |
Custom siRNA, 20nmol, CY3 Purity, RNAi is the best Way to Effectively Knock Down Gene Expression to Study Protein Function in a Wide Range of Cell Types, Use our GeneAssist Custom siRNA Builder to Design and Order a Custom siRNA | |
CY3 |