Learn More
Reichert™ Cuvets for Reichert Unistat™ Bilirubinometer
For use with Reichert UNISTAT Bilirubinometer (cat. no. AO10310C)
Brand:  Reichert™ 13102140
 View more versions of this product
Product Code. 12872003
  • 129.00 € / Pack of 51
Explore more special offers
Description
Description
  • For use with Unistat Bilirubinometer
  • Capacity: 20μL
Specifications
Specifications
Disposable Cuvette
Undiluted Serum
Reichert Unistat™ Bilirubinometer
20 μL