missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reichert™ Cuvets for Reichert Unistat™ Bilirubinometer
For use with Reichert UNISTAT Bilirubinometer (cat. no. AO10310C)
129.00 € - 742.00 €
Specifications
Sample Type | Undiluted Serum |
---|---|
For Use With (Equipment) | Reichert Unistat™ Bilirubinometer |
Capacity (Metric) | 20 μL |
Product Code | Brand | Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Price | Quantity & Availability | |||||
12918594
|
Reichert™
13103430 |
Low to Level Check Cuvette |
742.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12872003
|
Reichert™
13102140 |
Disposable Cuvette |
129.00 €
Pack of 51 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For use with Unistat Bilirubinometer
- Capacity: 20μL
Specifications
Undiluted Serum | |
Reichert Unistat™ Bilirubinometer | |
20 μL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Reichert™ Cuvets for Reichert Unistat™ Bilirubinometer