Learn More
Reichert™ Cuvets for Reichert Unistat™ Bilirubinometer
For use with Reichert UNISTAT Bilirubinometer (cat. no. AO10310C)
Brand:  Reichert™ 13103430
 View more versions of this product
Product Code. 12918594
  • 742.00 € / Each
Explore more special offers
Description
Description
  • Low-Level Check Cuvette
  • For use with Reichert Unistat Bilirubinometer
  • Capacity: 20μL
Specifications
Specifications
Low to Level Check Cuvette
Undiluted Serum
Reichert Unistat™ Bilirubinometer
20 μL