missing translation for 'onlineSavingsMsg'
Learn More
Cyanuric chloride, 99%, Thermo Scientific Chemicals Product Code.: 10665142

Cyanuric chloride, 99%, Thermo Scientific Chemicals

Product Code. 10665142
250 g, Glass bottle
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10665142

Brand: Thermo Scientific Acros 110892500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-77-0
Molecular Formula C3Cl3N3
Molecular Weight (g/mol) 184.40
MDL Number MFCD00006046
InChI Key MGNCLNQXLYJVJD-UHFFFAOYSA-N
Synonym cyanuric chloride, cyanuryl chloride, cyanuric acid chloride, s-triazine trichloride, trichlorocyanidine, cyanuric trichloride, tricyanogen chloride, sym-trichlorotriazine, cyanur chloride, trichloro-s-triazineShow MoreShow Less
PubChem CID 7954
ChEBI CHEBI:58964
SMILES ClC1=NC(Cl)=NC(Cl)=N1

Specifications

Melting Point 145.0°C to 148.0°C
Color Beige-Brown to Yellow
Density 1.9200g/mL
Boiling Point 190.0°C
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Quantity 250 g
Beilstein 26,35
Fieser 03,72; 04,522; 05,687; 06,149; 10,114; 15,105
Merck Index 152691
Specific Gravity 1.92
Solubility Information Solubility in water: reacts. Other solubilities: reacts with ethanol, 250g/L acetone (25°C), soluble in chloroform, hot ether, dioxane, ketones, and carbontetrachloride
IUPAC Name trichloro-1,3,5-triazine
Viscosity 0.57 mPa.s (150°C)
Formula Weight 184.41
Percent Purity 0.99
Physical Form Solidified Mass or Fragments
Chemical Name or Material Cyanuric chloride, 0.99
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468537 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number110890010,110890051,110892500
Lot NumberA0460357 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number110890010,110890051,110890050,110892500
Lot NumberA0447187 Certificate TypeCertificate of Analysis Date12/12/2023 Catalog Number110890010,110890051,110890050,110892500
Lot NumberA0441774 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number110890010,110890051,110890050,110892500
Lot NumberA0400937 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number110890010,110890051,110890050,110892500
compliance-icons compliance-icons
Product Identifier
  • Cyanuric chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
  • Acute toxicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyanuric chloride, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.