missing translation for 'onlineSavingsMsg'
Learn More

Contec™ Non-Sterile CyChlor
BIOPHARMA

Provided ready-to-use and is efficacious against bacteria and yeasts in 3 mins. Contec™ Non-Sterile CyChlor is a new broad spectrum disinfectant designed for every day use.

104.00 € - 118.00 €

Specifications

Form Liquid
pH 3 to 6
Sterility Non-sterile
Appearance Colorless
Autoclavable Not Autoclavable
View More Specs

Products 3
Product Code Brand Volume (Metric) Container Type Price Quantity & Availability  
Product Code Brand Volume (Metric) Container Type Price Quantity & Availability  
15929875
BIOPHARMA
View Documents
Contec™
FBC100CC
1 L Capped Container
104.00 €
6 bottles
Please to purchase this item. Need a web account? Register with us today!
15949875
BIOPHARMA
View Documents
Contec™
FBT100CC
1 L Trigger Spray Bottle
113.00 €
6 bottles
Please to purchase this item. Need a web account? Register with us today!
15939875
BIOPHARMA
View Documents
Contec™
FBC500CC
5 L Capped Container
118.00 €
2 bottles
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Broad spectrum efficacy in 3 mins saves time spent on biodecontamination.
  • Filtered to 0.2 microns and filled in a Grade A environment ensuring the product is free from contamination and particulates
  • Contains no quaternary ammonium or surfactant so it's very low residue, saving time on residue removal
  • No hazard classification so only basic PPE required and no special disposal is necessary
  • Trigger spray can be set to jet or spray - large droplet size reduces the risk of inhalation and provides good surface coverage
  • Double bagged packed in linear tear packaging so each bag is easy to open even when wearing gloves
  • Not classed as corrosive and can be used safely in all areas of the cleanroom — this needs a bit of random text deleting from the end
Specifications

Specifications

Liquid
Non-sterile
Not Autoclavable
Fluid
No
3 to 6
Colorless
Clear
Limited-use
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Contec™ Non-Sterile CyChlor

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.