missing translation for 'onlineSavingsMsg'
Learn More
Cyclobutanecarboxylic acid, 98%, Thermo Scientific Chemicals Product Code.: 10006093

Cyclobutanecarboxylic acid, 98%, Thermo Scientific Chemicals

Product Code. 10006093
100 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10006093

Brand: Thermo Scientific Acros 110901000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3721-95-7
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.12
MDL Number MFCD00001323
InChI Key TXWOGHSRPAYOML-UHFFFAOYSA-N
Synonym cyclobutylcarboxylic acid, cyclo-butyl formic acid, cyclobutane carboxylic acid, unii-0rdj7c51o0, pubchem14139, acmc-1cnqb, cyclobutan carboxylic acid, cyclobutyl carboxylic acid, cyclobutancecarboxylic acid, cyclobutane-carboxylic acidShow MoreShow Less
PubChem CID 19494
IUPAC Name cyclobutanecarboxylic acid
SMILES OC(=O)C1CCC1

Specifications

Melting Point -20.0°C
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 195.0°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C4H7CO2H
Refractive Index 1.4410 to 1.4450
Quantity 100 g
Beilstein 09, 5
Specific Gravity 1.04
Formula Weight 100.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclobutanecarboxylic acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR08J025 Certificate TypeCertificate of Analysis Date05/09/2023 Catalog Number110900250,110901000,110900050,11090081
Lot NumberA0420460 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number110900250,110901000,110900050,11090081
Lot NumberA0393838 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number110900250,110901000,110900050,11090081
Lot NumberA0413434 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number110900250,110901000,110900050,11090081
compliance-icons
Product Identifier
  • Cyclobutanecarboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclobutanecarboxylic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.