missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, ≥99.8%, Laboratory Reagent, Honeywell™
Laboratory Reagent,≥99.8%
78.23 € - 3535.07 €
Chemical Identifiers
CAS | 110-82-7 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.16 |
MDL Number | MFCD00003814 |
InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
Synonym | hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane |
PubChem CID | 8078 |
ChEBI | CHEBI:29005 |
IUPAC Name | cyclohexane |
SMILES | C1CCCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15627720
|
Honeywell Research Chemicals
C100307-1L |
1 L | Glass Bottle |
78.23 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15637720
|
Honeywell Research Chemicals
C100307-2.5L |
2.5 L | Glass Bottle |
156.26 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15677720
|
Honeywell Research Chemicals
C100307-5L |
5 L | Aluminum Bottle |
299.41 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15657720
|
Honeywell Research Chemicals
C100307-4X2.5L |
4 x 2.5 L | Glass Bottle |
522.92 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15607720
|
Honeywell Research Chemicals
C100307-10L |
10 L | Metal Can | ||||||
15667720
|
Honeywell Research Chemicals
C100307-4X5L |
4 x 5 L | Aluminum Bottle | ||||||
15647720
|
Honeywell Research Chemicals
C100307-25L |
25 L | Stainless Steel Can |
791.47 €
25 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15617720
|
Honeywell Research Chemicals
C100307-150KG |
150 kg | Metal Drum |
3535.07 €
150kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
110-82-7 | |
84.16 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
8078 | |
cyclohexane |
C6H12 | |
MFCD00003814 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
CHEBI:29005 | |
C1CCCCC1 |
Specifications
6.4°C | |
Colorless | |
81°C (1013 hPa) | |
500°F | |
Liquid | |
MFCD00003814 | |
UN1145 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
C1CCCCC1 | |
84.16 | |
CHEBI:29005 | |
≥99.8% |
110-82-7 | |
0.779g/mL at 25°C | |
0.09 | |
Characteristic | |
C6H12 | |
168.8mmHg (37.7°C), 77mmHg (20°C) | |
1900225 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
cyclohexane | |
8078 | |
84.16g/mol | |
Cyclohexane |
Safety and Handling
P210-P261-P273-P301 + P310-P331-P501
missing translation for 'dotInformation' : class 3 - PG 2 - Cyclohexane
missing translation for 'rtecsNumber' : 203-806-2
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Cyclohexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.