missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexane, CHROMASOLV™, for HPLC, ≥99.7%, Honeywell Riedel-de Haën™
CHROMASOLV™, for HPLC,≥99.7%
45.29 € - 1081.09 €
Chemical Identifiers
CAS | 110-82-7 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.16 |
MDL Number | MFCD00003814 |
InChI Key | XDTMQSROBMDMFD-UHFFFAOYSA-N |
Synonym | hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane |
PubChem CID | 8078 |
ChEBI | CHEBI:29005 |
IUPAC Name | cyclohexane |
SMILES | C1CCCCC1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15694530
|
Honeywell-Chromasolv
34855-100ML |
100 mL | Glass Bottle | ||||||
15604540
|
Honeywell-Chromasolv
34855-1L |
1 L | Glass Bottle |
137.19 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15624540
|
Honeywell-Chromasolv
34855-2L |
2 L | Glass Bottle | ||||||
15614540
|
Honeywell-Chromasolv
34855-2.5L |
2.5 L | Glass Bottle |
313.49 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15644540
|
Honeywell-Chromasolv
34855-6X1L |
6 x 1 L | Glass Bottle |
713.41 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15634540
|
Honeywell-Chromasolv
34855-4X2.5L |
4 x 2.5 L | Glass Bottle |
1081.09 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Chemical Identifiers
110-82-7 | |
84.16 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
8078 | |
cyclohexane |
C6H12 | |
MFCD00003814 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
CHEBI:29005 | |
C1CCCCC1 |
Specifications
4°C to 7°C (literature) | |
110-82-7 | |
C6H12 | |
MFCD00003814 | |
1900225 | |
XDTMQSROBMDMFD-UHFFFAOYSA-N | |
cyclohexane | |
8078 | |
≥99.7% | |
0.779 g/mL at 25 °C(lit.); 0.780 g/mL at 25 °C | |
−18°C |
max. 0.10 at 235 nm, max. 0.20 at 230 nm, max. 0.50 at 220 nm, max. 1.00 at 210 nm, max. 0.01 at 400 nm, max. 0.01 at 300 nm, max. 0.01 at 255 nm, max. 0.03 at 250 nm, max. 0.08 at 240 nm | |
≤100.0000% | |
C6H12 | |
UN1145 | |
hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane | |
C1CCCCC1 | |
84.16 | |
CHEBI:29005 | |
HPLC | |
81°C | |
Cyclohexane, CHROMASOLV™, for HPLC, ≥99.7% |
Safety and Handling
missing translation for 'shelfLife' : 1260 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Cyclohexane
missing translation for 'einecsNumber' : 203-806-2
Recommended Storage : Room Temp
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Cyclohexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.