missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, Extra Pure, SLR, Fisher Chemical™

44.40 € - 525.00 €

Specifications

CAS 110-82-7
Vapor Pressure 104mbar at 20°C
Viscosity 0.94 mPaS at 20°C
Vapor Density 2.9
Molecular Formula C6H12
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11458143
View Documents
Fisher Chemical
C/8920/PB08
500 mL Plastic Coated Glass bottle
44.40 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10625682
View Documents
Fisher Chemical
C/8920/PB17
2.5 L Plastic Coated Glass bottle
54.85 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10010100
View Documents
Fisher Chemical
C/8920/21
5 L HDPE Plastic Bottle
178.00 €
5 L
Please to purchase this item. Need a web account? Register with us today!
10275869
View Documents
Fisher Chemical
C/8920/FP21
5 L Plastic container
176.00 €
5 L
Please to purchase this item. Need a web account? Register with us today!
10487440
View Documents
Fisher Chemical
C/8920/25
25 L Metal can
525.00 €
25 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

110-82-7
0.94 mPaS at 20°C
C6H12
Liquid
6.5°C
84.15g/mol
104mbar at 20°C
2.9
3814
Colorless
81°C
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cyclohexane, Extra Pure, SLR, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.