Learn More
Cyclohexane, Extra Pure, SLR, Fisher Chemical™
Brand: Fisher Chemical
75.87 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description

Specifications
110-82-7 | |
5 L | |
0.94 mPaS at 20°C | |
C6H12 | |
Liquid | |
6.5°C | |
84.15g/mol |
HDPE Plastic Bottle | |
104mbar at 20°C | |
2.9 | |
3814 | |
Colorless | |
81°C |
Safety and Handling
- Cyclohexane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Aspiration toxicity Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.