missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals
45.53 € - 625.11 €
Chemical Identifiers
CAS | 87100-15-0 |
---|---|
Molecular Formula | C12H23BO2 |
Molecular Weight (g/mol) | 210.124 |
MDL Number | MFCD04038749 |
InChI Key | OUEVCDGYTKLNMJ-UHFFFAOYSA-N |
Synonym | cyclohexylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-cyclohexyl-4,4,5,5-tetramethyl, cyclohexylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane, pubchem7924, cyclohexylboronic acid, pinacol eser, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane |
PubChem CID | 3668596 |
IUPAC Name | 2-cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2CCCCC2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15441269
|
Thermo Scientific Alfa Aesar
H64178.03 |
1 g |
45.53 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15451269
|
Thermo Scientific Alfa Aesar
H64178.06 |
5 g |
153.77 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15461269
|
Thermo Scientific Alfa Aesar
H64178.14 |
25 g |
625.11 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
87100-15-0 | |
210.124 | |
OUEVCDGYTKLNMJ-UHFFFAOYSA-N | |
3668596 | |
B1(OC(C(O1)(C)C)(C)C)C2CCCCC2 |
C12H23BO2 | |
MFCD04038749 | |
cyclohexylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-cyclohexyl-4,4,5,5-tetramethyl, cyclohexylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane, pubchem7924, cyclohexylboronic acid, pinacol eser, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane | |
2-cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane |
Specifications
87100-15-0 | |
C12H23BO2 | |
cyclohexylboronic acid pinacol ester, 1,3,2-dioxaborolane, 2-cyclohexyl-4,4,5,5-tetramethyl, cyclohexylboronic acid, pinacol ester, 4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane, pubchem7924, cyclohexylboronic acid, pinacol eser, 1-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl cyclohexane | |
B1(OC(C(O1)(C)C)(C)C)C2CCCCC2 | |
210.124 | |
210.12 | |
Liquid |
Colorless | |
MFCD04038749 | |
OUEVCDGYTKLNMJ-UHFFFAOYSA-N | |
2-cyclohexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane | |
3668596 | |
97% | |
Cyclohexylboronic acid pinacol ester |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only