Learn More
Cyclopentylacetyl chloride, 97%, Thermo Scientific Chemicals
79.80 € - 307.53 €
Chemische Identifikatoren
CAS | 1122-99-2 |
---|---|
Molecular Formula | C7H11ClO |
Molecular Weight (g/mol) | 146.61 |
MDL Number | MFCD00236031 |
InChI Key | NILLIUYSJFTTRH-UHFFFAOYSA-N |
Synonym | cyclopentylacetyl chloride, cyclopentaneacetyl chloride, cyclopentylacetylchloride, cylopentylacetyl chloride, acmc-1brgh, cyclopentaneacetylchloride, cyclopentylacetic chloride, cyclopentanacetyl chloride, cyclopentyl acetyl chloride, cyclopentyl-acetyl chloride |
PubChem CID | 3485182 |
IUPAC Name | 2-cyclopentylacetyl chloride |
SMILES | ClC(=O)CC1CCCC1 |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11393586
|
Thermo Scientific Alfa Aesar
L14562.06 |
5 g |
79.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11303596
|
Thermo Scientific Alfa Aesar
L14562.14 |
25 g |
307.53 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Cyclopentylacetyl chloride is used as a pharmaceutical intermediate and in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCyclopentylacetyl chloride is used as a pharmaceutical intermediate and in chemical research.
Solubility
Reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to moisture. Incompatible with bases and oxidizing agents. Store under inert gas.
Chemische Identifikatoren
1122-99-2 | |
146.61 | |
NILLIUYSJFTTRH-UHFFFAOYSA-N | |
3485182 | |
ClC(=O)CC1CCCC1 |
C7H11ClO | |
MFCD00236031 | |
cyclopentylacetyl chloride, cyclopentaneacetyl chloride, cyclopentylacetylchloride, cylopentylacetyl chloride, acmc-1brgh, cyclopentaneacetylchloride, cyclopentylacetic chloride, cyclopentanacetyl chloride, cyclopentyl acetyl chloride, cyclopentyl-acetyl chloride | |
2-cyclopentylacetyl chloride |
Spezifikation
1122-99-2 | |
186°C | |
Pungent | |
1.4625 | |
UN3265 | |
Moisture sensitive | |
Reacts with water. | |
ClC(=O)CC1CCCC1 | |
146.61 | |
146.62 | |
Cyclopentylacetyl chloride |
1.087 | |
71°C (160°F) | |
C7H11ClO | |
MFCD00236031 | |
1906113 | |
cyclopentylacetyl chloride, cyclopentaneacetyl chloride, cyclopentylacetylchloride, cylopentylacetyl chloride, acmc-1brgh, cyclopentaneacetylchloride, cyclopentylacetic chloride, cyclopentanacetyl chloride, cyclopentyl acetyl chloride, cyclopentyl-acetyl chloride | |
NILLIUYSJFTTRH-UHFFFAOYSA-N | |
2-cyclopentylacetyl chloride | |
3485182 | |
97% |
Sicherheit und Handhabung
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.