Learn More
Sartorius™ Cylindrical Calibration Weight
Confidently calibrate your scale with this finely crafted, special steel weight. Sartorius™ Cylindrical Calibration Weight is highly polished and non-magnetizable.
Brand: Sartorius™ YCW624-AC-00
This item is not returnable.
View return policy
Description
- Cylindrical weight
- Special steel
- Highly polished
- Non-magnetizable
- Highly corrosion-resistant
- Class F2 - suitable for use for traceability of OIML class M1 weights and lower
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Calibration Weight | |
Special Steel | |
7,950kg/m3 | |
Laboratory scale |
Class F2 - suitable for use for traceability of OIML class M1 weights and lower | |
Plastic | |
2 kg |