missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Cylindrical Calibration Weight

Confidently calibrate your scale with this finely crafted, special steel weight. Sartorius™ Cylindrical Calibration Weight is highly polished and non-magnetizable.

217.00 € - 223.00 €

Specifications

Product Type Calibration Weight
Classification Class F2 - suitable for use for traceability of OIML class M1 weights and lower
Material Special Steel
Case Material Plastic
Density 7,950kg/m3
View More Specs

Products 2
Product Code Brand Mass Price Quantity & Availability  
Product Code Brand Mass Price Quantity & Availability  
15336660
View Documents
Sartorius™
YCW5248-02
200 g
217.00 €
200g
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
11363424
View Documents
Sartorius™
YCW624-AC-00
2 kg
223.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • Cylindrical weight
  • Special steel
  • Highly polished
  • Non-magnetizable
  • Highly corrosion-resistant
  • Class F2 - suitable for use for traceability of OIML class M1 weights and lower
Specifications

Specifications

Calibration Weight
Special Steel
7,950kg/m3
Class F2 - suitable for use for traceability of OIML class M1 weights and lower
Plastic
Laboratory scale
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.