missing translation for 'onlineSavingsMsg'
Learn More
Cystamine dihydrochloride, 97+%, Thermo Scientific Chemicals Product Code.: 11409927

Cystamine dihydrochloride, 97+%, Thermo Scientific Chemicals

Product Code. 11409927
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11409927

Brand: Thermo Scientific Alfa Aesar B22873.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cystamine dihydrochloride acts as an anti-infective agent, which is used in the treatment of urinary tract infections. It is also used as a radiation-protective agent that interferes with sulfhydryl enzymes. Further, it serves as a heparin antagonist and sulfhydryl modifying reagent. In addition to this, it is used as an inhibitor of TGase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cystamine dihydrochloride acts as an anti-infective agent, which is used in the treatment of urinary tract infections. It is also used as a radiation-protective agent that interferes with sulfhydryl enzymes. Further, it serves as a heparin antagonist and sulfhydryl modifying reagent. In addition to this, it is used as an inhibitor of TGase.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-17-7
Molecular Formula C4H14Cl2N2S2
Molecular Weight (g/mol) 225.19
MDL Number MFCD00012905
InChI Key YUFRRMZSSPQMOS-UHFFFAOYSA-N
Synonym cystamine dihydrochloride, cystamin dihydrochloride, ethanamine, 2,2'-dithiobis-, dihydrochloride, usaf cb-34, 2,2'-disulfanediyldiethanamine dihydrochloride, 2-aminoethyl disulfide dihydrochloride, bis 2-aminoethyl disulfide dihydrochloride, cystamine 2hcl, 2,2'-dithiodi ethylammonium dichloride, decarboxycystine dihydrochlorideShow MoreShow Less
PubChem CID 5941
IUPAC Name 2-(2-aminoethyldisulfanyl)ethanamine;dihydrochloride
SMILES C(CSSCCN)N.Cl.Cl

Specifications

Melting Point 217°C to 220°C
Assay Percent Range 98+%
Odor Odorless
Linear Formula (H2NCH2CH2S)2·2HCl
Quantity 100 g
Beilstein 3616850
Sensitivity Hygroscopic
Merck Index 14,2776
Solubility Information Soluble in water.
Formula Weight 225.2
Percent Purity ≥97%
Chemical Name or Material Cystamine dihydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10247323 Certificate TypeCertificate of Analysis Date13/07/2023 Catalog NumberB22873.22,B22873,B22873.14
Lot Number10245103 Certificate TypeCertificate of Analysis Date30/12/2022 Catalog NumberB22873.22,B22873.14
Lot NumberJ1972A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22873.22,B22873.14
Lot Number10234234 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22873.22,B22873.14
Lot NumberC22W015 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22873.22,B22873.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cystamine dihydrochloride, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.