Learn More
Epredia™ Cytospin™ 4 Cytofunnel™ Starter Kit
Accessory Starter Kit for the Epredia™ Cytospin™ 4 Cytocentrifuge. (Centrifuge is sold separately, Cat. No. A78300003)
Brand:  Epredia™ 3120110
 View more versions of this product
Product Code. 12017159
  • 2240.00 € / Each
Explore more special offers
Description
Description
The Cytofunnel Starter Kit provides everything needed for new Cytospin 4 users.
Available in both standard clipped Cytofunnel and EZ Cytofunnel options:
  • Clipped Cytofunnel Starter Kit
    • Includes: 50 Single White Cytofunnels, 25 Double Cytofunnels, Cytoslides, 12 Cytoclips
  • EZ Cytofunnel Starter Kit
    • Includes: 25 EZ Megafunnels, 20 EZ Single White Cytofunnels, 20 EZ Double Cytofunnels and Cytoslides
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytospin™ 4 Cytocentrifuge
Clipped Funnel Starter Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Epredia™ Cytospin™ 4 Cytofunnel™ Starter Kit > Clipped funnel starter kit