missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Fructose, Pure, Fisher Chemical™

13.40 € - 26.00 €

Chemical Identifiers

CAS 57-48-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number 148910
InChI Key BJHIKXHVCXFQLS-UYFOZJQFSA-N
Synonym d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
PubChem CID 5984
ChEBI CHEBI:48095
IUPAC Name (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
SMILES C(C(C(C(C(=O)CO)O)O)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11482614
View Documents
Fisher Chemical
F/1950/48
100 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10346040
View Documents
Fisher Chemical
F/1950/50
250 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

D(-)-Fructose, Pure

Specifications

Chemical Identifiers

57-48-7
180.156
BJHIKXHVCXFQLS-UYFOZJQFSA-N
5984
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
C6H12O6
148910
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
CHEBI:48095
C(C(C(C(C(=O)CO)O)O)O)O

Specifications

57-48-7
White
Plastic powder jar
148910
BJHIKXHVCXFQLS-UYFOZJQFSA-N
(3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one
5984
180.16g/mol
103°C
5 to 7
C6H12O6
d---fructose, d--fructose, 3s,4r,5r-1,3,4,5,6-pentahydroxyhexan-2-one, furucton, arabino-hexulose, keto-d-fructose, d-levulose, sugar, fruit, fructose, d, krystar 300
C(C(C(C(C(=O)CO)O)O)O)O
180.156
CHEBI:48095
Solid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • D(-)-Fructose

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.