missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals D-Lysine hydrochloride, 99+% Product Code.: 10308520

Thermo Scientific Chemicals D-Lysine hydrochloride, 99+%

Product Code. 10308520
10 g, Glass Bottle
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass Bottle
This item is not returnable. View return policy

Product Code. 10308520

Brand: Thermo Scientific Chemicals 243080100

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7274-88-6
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.65
MDL Number MFCD00012920
InChI Key BVHLGVCQOALMSV-XZNNNFJINA-N
Synonym d-lysine hydrochloride, d-lysine monohydrochloride, d-lysine hcl, d-lysine, monohydrochloride, h-d-lys-oh.hcl, d-lysine, hydrochloride, unii-kk611jde1n, lysine, monohydrochloride, d, r-2,6-diaminohexanoic acid hydrochloride, lysine, hydrochloride, dShow MoreShow Less
PubChem CID 81691
ChEBI CHEBI:53558
SMILES Cl.NCCCC[C@@H](N)C(O)=O

Specifications

Arsenic (As) 1ppm max.
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 30ppm max.
Assay Percent Range 99+%
Loss on Drying 0.3% max. (105°C, 3 hrs)
Packaging Glass Bottle
Linear Formula H2N(CH2)4CH(NH2)COOH·HCl
Specific Rotation -20.5° to -21.5° (20°C, 589nm) (c=8, 6N HCl)
Fieser 01,959; 07,79; 01,977
Sulfated Ash 0.1% max.
IUPAC Name (2R)-2,6-diaminohexanoic acid;hydrochloride
Specific Rotation Condition −21° (20°C c=8,6N HCl)
Quantity 10 g
Formula Weight 182.65
Percent Purity 99.0 to 101.0%
Physical Form Crystalline Powder
Chemical Name or Material D-Lysine hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453983 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number243080500,243080010,243080100
Lot NumberA0402569 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number243080500,243080010,243080100
Lot NumberA0439863 Certificate TypeCertificate of Analysis Date24/03/2022 Catalog Number243080500,243080010,243080100
Lot NumberA0410763 Certificate TypeCertificate of Analysis Date07/02/2022 Catalog Number243080500,243080010,243080100
Product Identifier
  • D-Lysine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals D-Lysine hydrochloride, 99+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.